Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYB5R2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179540
Description
CYB5R2 Polyclonal specifically detects CYB5R2 in Rat samples. It is validated for Western Blot.Specifications
CYB5R2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
B5R.2, cytochrome b5 reductase 2 | |
Rabbit | |
30 kDa | |
100 μL | |
Cardiovascular Biology | |
51700 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_001014266 | |
CYB5R2 | |
Synthetic peptide directed towards the C terminal of human Cyb5r2The immunogen for this antibody is Cyb5r2. Peptide sequence WEYSSGFITADMIKEHLPPPGEATLILVCGPPPLIQEAAHPSLEQLGYTK. | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Horse: 85%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction