Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYB5R2 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | CYB5R2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179540
|
Novus Biologicals
NBP179540 |
100 μL |
Each of 1 for $436.00
|
|
Description
CYB5R2 Polyclonal specifically detects CYB5R2 in Rat samples. It is validated for Western Blot.Specifications
CYB5R2 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
B5R.2, cytochrome b5 reductase 2 | |
CYB5R2 | |
IgG | |
Affinity Purified | |
30 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_001014266 | |
51700 | |
Synthetic peptide directed towards the C terminal of human Cyb5r2The immunogen for this antibody is Cyb5r2. Peptide sequence WEYSSGFITADMIKEHLPPPGEATLILVCGPPPLIQEAAHPSLEQLGYTK. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title