Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYB5R3 Antibody (OTI2A10), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270572APCCY7
Description
CYB5R3 Monoclonal specifically detects CYB5R3 in Human, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.Specifications
CYB5R3 | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
CYB5R3 | |
Full length recombinant protein of human CYB5R3 (NP_000389) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Canine, Primate, Monkey | |
IgG2b |
Flow Cytometry | |
OTI2A10 | |
Flow Cytometry | |
B5RNADH-cytochrome b5 reductase 3, Cytochrome b5 reductase, cytochrome b5 reductase 3, DIA1NADH-cytochrome b5 reductase 3 membrane-bound form, diaphorase (NADH) (cytochrome b-5 reductase), Diaphorase-1, EC 1.6.2.2, NADH-cytochrome b5 reductase 3 soluble form | |
Mouse | |
Affinity Purified | |
Cancer, Lipid and Metabolism | |
1727.0 | |
Store at 4°C in the dark. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction