Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYB5R3 Mouse anti-Human, Canine, Monkey, PE/Atto594, Clone: OTI2A10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270572PEATT594

Catalog No. N270572P594


Add to cart

Description

Description

CYB5R3 Monoclonal antibody specifically detects CYB5R3 in Human, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

CYB5R3
OTI2A10
Flow Cytometry
Affinity Purified, Conjugated
CYB5R3
Full length recombinant protein of human CYB5R3 (NP_000389) produced in HEK293T cells.
Immunogen affinity purified
Cancer, Lipid and Metabolism
Primary
1727.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
B5RNADH-cytochrome b5 reductase 3, Cytochrome b5 reductase, cytochrome b5 reductase 3, DIA1NADH-cytochrome b5 reductase 3 membrane-bound form, diaphorase (NADH) (cytochrome b-5 reductase), Diaphorase-1, EC 1.6.2.2, NADH-cytochrome b5 reductase 3 soluble form
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit