Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYBRD1 Polyclonal Antibody
CYBRD1 Polyclonal Antibody
Supplier: Thermo Scientific PA566036
Description
CYBRD1 Polyclonal Antibody for ICC/IF

Specifications
CYBRD1 | |
Polyclonal | |
Unconjugated | |
CYBRD1 | |
2210407P13Rik; CYB561A2; CYBRD1; Cytochrome b reductase 1; cytochrome b561 family, member A2; Dcytb; Duodenal cytochrome b; ferric-chelate reductase 3; FRRS3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79901 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q53TN4 | |
CYBRD1 | |
Recombinant Human CYBRD1. Recombinant protein control fragment (Product #RP-106726). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction