Learn More
Selleck Chemical LLCSupplier Diversity Partner CYC116 200mg 693228-63-6  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S1171200MG
CYC116 is a potent inhibitor of Aurora A/B with Ki of 8.0 nM/9.2 nM, is less potent to VEGFR2 (Ki of 44 nM), with 50-fold greater potency than CDKs, not active against PKA, Akt/PKB, PKC, no effect on GSK-3α/β, CK2, Plk1 and SAPK2A. Phase 1.
Catalog No. 50-689-0