Learn More
Enzo Life SciencesSupplier Diversity Partner cAMP Complete ELISA kit (1x96wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI900163
Most sensitive (0.037 pmol/ml) and complete colorimetric kit for intra- and extracellular cAMP quantification. Alternative Name: cyclic AMP. Sensitivity: Assay Buffer : 0.30 pmol/ml (non-acetylated) 0.039 pmol/ml (acetylated) HCl: 0.39 pmol/ml (non-acetylated) 0.037 pmol/ml (acetylated) (range 0.78 - 200 pmol/ml (non-acetylated); 0.078 - 20 pmol/ml (acetylated)). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR IgG Microtiter plate, Conjugate, Antibody, 0.1M HCl, Assay buffer 2, Neutralizing reagent, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Triethylamine, Acetic anhydride
Catalog No. 50-658-189