Learn More
Enzo Life SciencesSupplier Diversity Partner Direct cAMP ELISA kit (5x96wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI901066
Most sensitive (0.037 pmol/ml) colorimetric kit for intracellular cAMP quantification. Alternative Name: cyclic AMP. Sensitivity: (non-acetylated) 0.39 pmol/ml; (acetylated) 0.037 pmol/ml (range (non-acetylated) 0.78 - 200 pmol/ml;(acetylated) 0.078 - 20 pmol/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR Microtiter plate, Conjugate, Antibody, 0.1M HCl, Neutralizing reagent, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Acetic anhydride, Triethylamine
Catalog No. 50-658-219