Learn More
Enzo Life SciencesSupplier Diversity Partner cAMP ELISA kit (1x96wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI900067
Most sensitive (0.039 pmol/ml) colorimetric kit for extracellular cAMP quantification. This is a colorimetric competitive enzyme immunoassay kit for the quantitative determination of extracellular cAMP. Sensitivity is increased 10-fold by acetylation (reagents included). Absorbance is read at 405 nm. The ready-to-use liquid color-coded reagents provided reduce error. Alternative Name: cyclic AMP. Sensitivity: (non-acetylated) 0.30 pmol/ml; (acetylated) 0.039 pmol/ml (range (non-acetylated) 0.78 - 200 pmol/ml;(acetylated) 0.078 - 20 pmol/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer 2, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Acetic anhydride, Triethylamine
Catalog No. 50-658-125