Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin A Mouse anti-Human, Clone: 1, Invitrogen™

Catalog No. PIMA529090
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA529090 100 μL
1 options

Catalog No. PIMA529090

Supplier: Thermo Scientific MA529090

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).

Cyclins are regulatory subunits of the cyclin-dependent kinases (cdk′s). They are expressed temporally to control transition at different specific phases of the cell cycle. In mammalian somatic cells, cyclin A is required for S-phase and passage through G2-phase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin A
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CCNA1
Gene Accession No. P78396
Gene Alias cyclin-A1
Gene Symbols CCNA1
Host Species Mouse
Immunogen Recombit Human Cyclin A1/CCNA1 protein
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8900
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.