Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin A1 Mouse anti-Xenopus, Clone: XLA1-1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30782910UG

 View more versions of this product

Catalog No. NB123131


Add to cart

Description

Description

Cyclin A1 Monoclonal antibody specifically detects Cyclin A1 in Xenopus samples. It is validated for Western Blot
Specifications

Specifications

Cyclin A1
XLA1-1
Unconjugated
10 mM PBS with 0.05% BSA
cyclin A1, cyclin-A1
Recombinant full-length Xenopus laevis Cyclin A1 protein
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Xenopus
Western Blot
0.2 mg/ml
Western Blot 1-2 ug/ml
Purified
Mouse
IgG1 Kappa
100 μg
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors
Primary
8900
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit