Learn More
Cyclin A2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 182021AP
Catalog No. 50-173-2044
Add to cart
Description
Description
DCP1B is a core component of the mRNA decapping complex, a key factor in the regulation of mRNA decay.
Specifications
Specifications
Cyclin A2
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P20248, P51943
CCN1, CCNA, CCNA2, Cyclin A, cyclin A2
Rabbit
IgG
150 μL
-20°C
Primary
12428, 890
Immunoprecipitation, Western Blot
Unconjugated
CCNA2
Liquid
CCNA2
Cyclin A2 Fusion Protein Ag12765
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse