Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cyclin B1 Ab-4, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and immunoprecipitation experiments.

Specifications

Antigen Cyclin B1 Ab-4
Clone GNS11
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Host Species Murine
View More Specs

Products 2
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS869P1
View Documents
Thermo Scientific™
MS869P1
Purified Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS869R1
View Documents
Thermo Scientific™
MS869R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cyclin B1 Ab-4, Mouse Monoclonal Antibody.

In mammals, cyclin B associates with inactive p34cdc2. Cyclin B-p34cdc2 plays a critical role in G2 to M transition.

Host Species: Mouse

Clone: GNS11

Isotype: IgG2a

Species Reactivity: Human, Mouse, Rat and Hamster. Others not known.

Epitope: aa 1-306

Immunogen: Human cyclin B1 protein

Molecular Weight: 62 kDa

Positive Control: LS174T cells, Tonsil or breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cyclin B1 Ab-4
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Murine
Cell Cycle
Human cyclin B1 protein
GNS11
Monoclonal
RUO
Primary
IgG2a
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit