Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin B1 Mouse anti-Human, DyLight 755, Clone: 238, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243612IR

Catalog No. NBP243612IR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Cyclin B1
238
Western Blot
Purified
CCNB1
Recombinant protein encompassing a sequence within the center region of human Cyclin B1. The exact sequence is proprietary.
Protein G purified
Cancer, Cell Biology, Cell Cycle and Replication, Mitotic Regulators, Phospho Specific, Tumor Suppressors
Primary
891
Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit