Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cyclin B1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting and flow cytometry applications with Thermo Scientific™ Cyclin B1, Mouse Monoclonal Antibody.

Specifications

Antigen Cyclin B1
Clone V152
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS338R1
View Documents
Thermo Scientific™
MS338R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting and flow cytometry applications with Thermo Scientific™ Cyclin B1, Mouse Monoclonal Antibody.

In mammals, cyclin B associates with inactive p34cdc2. Cyclin Bp34cdc2 plays a critical role in G2 to M transition.

Host Species: Mouse

Clone: V152

Isotype: IgG1

Species Reactivity: Human, Mouse, Rat and Hamster. Others not known.

Epitope: Not determined

Immunogen: Hamster cyclin B1 protein

Molecular Weight: 62 kDa

Positive Control: Jurkat cells, Tonsil or breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Not suitable)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cyclin B1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RUO
Primary
IgG1
V152
Monoclonal
Murine
Cell Cycle
Hamster cyclin B1 protein
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit