Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin B2 Mouse anti-Mammalia, Xenopus, Allophycocyanin/Cy7, Clone: X29.2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB1002649APCCY7

Catalog No. N102649APC7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Cyclin B2 Monoclonal antibody specifically detects Cyclin B2 in Mammalia, Xenopus samples. It is validated for Flow Cytometry.
Specifications

Specifications

Cyclin B2
X29.2
Flow Cytometry
Conjugated, Purified
CCNB2
Xenopus laevis cyclin B2
Protein A or G purified
Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary
9133.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
cyclin B2, G2/mitotic-specific cyclin-B2, HsT17299
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Mammalia, Xenopus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit