Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cyclin D1/Bcl-1 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation, biological blockade and flow cytometry applications.

Manufacturer:  Thermo Scientific™ MS210P1

 View more versions of this product

Catalog No. MS210P1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation, biological blockade and flow cytometry applications with Thermo Scientific™ Cyclin D1/Bcl-1 Ab-1, Mouse Monoclonal Antibody.

Cyclin D1or PRAD-1 or bcl-1 is one of the key cell cycle regulators and is a putative proto-oncogene overexpressed in a wide variety of human neoplasms including mantle cell lymphomas (MCL)

Host Species: Mouse

Clone: DCS-6

Isotype: IgG2a/κ

Species Reactivity: Human, Monkey, Mouse, Rat and Dog.

Epitope: Not determined

Immunogen: Human recombinant full-length cyclin D1 protein

Molecular Weight: 36 kDa

Positive Control: MAD109 cells; Mantle cell lymphomas and breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Not Suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Biological Blockade
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cyclin D1/Bcl-1 Ab-1
Monoclonal
200μg/mL
Concentrated
Human recombinant full length cyclin D1 protein
500μL
Cell Cycle
Primary
Canine, Human, Murine, Non-human Primate, Rat
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
DCS-6
Unconjugated
Murine
IgG2a kappa
RUO
Primary
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit