Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D1 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI1G2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270244AF405

Catalog No. N270244A405


Add to cart

Description

Description

Cyclin D1 Monoclonal antibody specifically detects Cyclin D1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Cyclin D1
OTI1G2
Western Blot
Affinity Purified, Conjugated
CCND1
Full length recombinant protein of human Cyclin D1 (NP_444284) produced in E.coli.
Immunogen affinity purified
Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers, Wnt Signaling Pathway
Primary
595.0
Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit