Learn More
Cyclin D1 Mouse anti-Human, Mouse, Rat, Primate, Alexa Fluor™ 532, Clone: DCS-6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233138AF532
Catalog No. NP233138A32
Add to cart
Description
Description
Cyclin D1 Monoclonal antibody specifically detects Cyclin D1 in Human,Mouse,Rat,Primate samples. It is validated for ELISA.
Specifications
Specifications
Cyclin D1
DCS-6
ELISA
B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31
Mouse
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Recognizes a protein of 36kDa, identified as cyclin D1. Cyclin D1, one of the key cell cycle regulators, is a putative proto-oncogene overexpressed in a wide variety of human neoplasms. This antibody neutralizes the activity of cyclin D1 in vivo. About 60% of mantle cell lymphomas (MCL) contain a t(11; 14)(q13; q32) translocation resulting in over-expression of cyclin D1. This antibody is useful in identifying mantle cell lymphomas (cyclin D1 positive) from CLL/SLL and follicular lymphomas (cyclin D1 negative). Occasionally, hairy cell leukemia and plasma cell myeloma weakly express Cyclin D1.
ELISA
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CCND1
Human recombinant full length Cyclin D1 protein was used as immunogen to generate this antibody.
0.1 mL
Primary
595
Human, Mouse, Primate, Rat