Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cyclin D2 Ab-2, Mouse Monoclonal Antibody

Ensure accurate and reproducible results in western blotting, immunofluorescence, immunoprecipitation, biological blockade and flow cytometry experiments.

$221.50 - $742.50

Specifications

Antigen Cyclin D2 Ab-2
Clone DCS-3.1
Applications Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
MS213P
View Documents
Thermo Scientific™
MS213P
200μg/mL 1mL Each for $732.50
Add to cart
 
MS213P0
View Documents
Thermo Scientific™
MS213P0
200μg/mL 100μL Each for $221.50
Add to cart
 
MS213P1
View Documents
Thermo Scientific™
MS213P1
200μg/mL 500μL Each for $476.50
Add to cart
 
MS213P1ABX
View Documents
Thermo Scientific™
MS213P1ABX
1mg/mL 100μL Each for $508.50
Add to cart
 
MS213PABX
View Documents
Thermo Scientific™
MS213PABX
1mg/mL 200μL Each for $742.50
Add to cart
 
Description

Description

Ensure accurate and reproducible results in western blotting, immunofluorescence, immunoprecipitation, biological blockade and flow cytometry experiments with Thermo Scientific™ Cyclin D2 Ab-2, Mouse Monoclonal Antibody.

Cyclin D2 is a G1 cyclin required for G1-phase progression and is a strong candidate for a proto-oncogene. Cyclin D2 can phosphorylate pRB when associated with cdk4 and/or cdk6.

Host Species: Mouse

Clone: DCS-3.1

Isotype: IgG2a/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified human recombinant full-length cyclin D2 protein

Molecular Weight: 34 kDa

Positive Control: U-2-OS, C82, RD, Bristol-8 or LoVo36 cells. Note that T47D, ZR75, SKBR3, MCF-7, BT-20, MDA468, HBL100, BT5549, HeLa, Saos2 and HT29 cells are negative for cyclin D2.

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Biological Blockade
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cyclin D2 Ab-2
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
Murine
Cell Cycle
Purified human recombinant full length cyclin D2 protein
DCS-3.1
Monoclonal
Concentrated
RUO
Primary
IgG2a kappa
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit