Learn More
Cyclin D2 Antibody (OTI5A6), DyLight 650, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270344C
Catalog No. NBP270344C
Add to Cart
Description
Description
Cyclin D2 Monoclonal specifically detects Cyclin D2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
Cyclin D2
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
CCND2
Full length recombinant protein of human Cyclin D2 (NP_001750) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG2b
Western Blot
OTI5A6
Western Blot
cyclin D2, G1/S-specific cyclin D2, G1/S-specific cyclin-D2, KIAK0002, MGC102758
Mouse
Affinity Purified
Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Wnt Signaling Pathway
894.0
Store at 4°C in the dark.
Recommendations
Recommendations