Learn More
Cyclin D2 Mouse anti-Human, Unconjugated, Clone: 1A12A8, Proteintech
Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 670481IG150UL
Description
The Cyclin D2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Cyclin D2. This antibody recognizes human, mouse antigen. The Cyclin D2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Cyclin D2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse |
Western Blot, ELISA | |
1A12A8 | |
Unconjugated | |
150 μL | |
894 | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.