Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cyclin D3 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry applications.

$189.10 - $757.50

Specifications

Antigen Cyclin D3 Ab-1
Clone DCS-22
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS215P
View Documents
Thermo Scientific™
MS215P
Purified Ab with BSA and Azide 1mL Each for $701.00
Add to cart
 
MS215P0
View Documents
Thermo Scientific™
MS215P0
Purified Ab with BSA and Azide 100μL Each for $189.10
Add to cart
 
MS215P1
View Documents
Thermo Scientific™
MS215P1
Purified Ab with BSA and Azide 500μL Each for $570.00
Add to cart
 
MS215P1ABX
View Documents
Thermo Scientific™
MS215P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $508.50
Add to cart
 
MS215PABX
View Documents
Thermo Scientific™
MS215PABX
Ab without BSA/Azide (Frozen) 200μL Each for $757.50
Add to cart
 
MS215R7
View Documents
Thermo Scientific™
MS215R7
Ready-to-use for IHC (UVLP) 7mL Each for $417.00
Add to cart
 
MS215R1
View Documents
Thermo Scientific™
MS215R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry applications with Thermo Scientific™ Cyclin D3 Ab-1, Mouse Monoclonal Antibody.

Cyclin D3 is a G1 cyclin closely related to cyclin D1 and D2. Cyclins D1, D2, and D3 form active kinase complexes in vivo with a distinct subset of cdks, particularly with cdk4 and cdk6.

Host Species: Mouse

Clone: DCS-22

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified human recombinant full-length cyclin D3 protein

Molecular Weight: 34 kDa

Positive Control: LS174T cells, Breast CA or tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native only)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cyclin D3 Ab-1
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RUO
Primary
IgG2a, κ
DCS-22
Monoclonal
Murine
Cell Cycle
Purified human recombinant full length cyclin D3 protein
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit