Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D3 Rabbit anti-Human, Mouse, Clone: SD20-43, Invitrogen™

Rabbit Monoclonal Antibody

Manufacturer:  Thermo Scientific MA532366

Catalog No. PIMA532366


Add to cart

Description

Description

Rabbit Monoclonal Antibody

Cyclin D3 Monoclonal antibody specifically detects Cyclin D3 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Specifications

Specifications

Cyclin D3
Monoclonal
1 mg/mL
TBS with 1% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
P30281, P30282
9230106B05Rik; AA682053; AL024085; AW146355; C78795; CCND 3; Ccnd3; CGD3; cyclin D3; CyclinD3; Cyl-3; D3 type cyclin; G1/S-specific cyclin-D3; OTTHUMP00000223415; RP5-973N23.3
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
12445, 896
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
SD20-43
Unconjugated
CCND3
Liquid
CCND3
C terminal human Cyclin D3
Protein A
RUO
Antibody
Monoclonal
Human, Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit