Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cyclin E Ab-2, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and flow cytometry experiments.

Specifications

Antigen Cyclin E Ab-2
Clone HE12
Applications Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Format Concentrated
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS870P1
View Documents
Thermo Scientific™
MS870P1
Purified Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and flow cytometry experiments with Thermo Scientific™ Cyclin E Ab-2, Mouse Monoclonal Antibody.

Cyclin E is a regulatory subunit of cdk-2 and controls G1/S transition during the mammalian cell cycle. Multiple isoforms of cyclin E are expressed only in tumors but not in the normal tissues. Qualitative and quantitative alterations in cyclin E protein have been implicated as indicators of worse prognosis in various cancers.

Host Species: Mouse

Clone: HE12

Isotype: IgG1

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant human cyclin E protein

Molecular Weight: Doublet of 50 kDa and a single band of 42 kDa

Positive Control: SKBR3 or LS174T cells; Breast CA or placenta

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Not suitable)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cyclin E Ab-2
Flow Cytometry, Immunofluorescence, Western Blot
Concentrated
RUO
Primary
IgG1
HE12
Monoclonal
Murine
Cell Cycle
Recombinant human cyclin E protein
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit