Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin E1 Mouse anti-Human, Clone: CCNE1/2460, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30748420UG
Description
Cyclin E1 Monoclonal specifically detects Cyclin E1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.Specifications
Cyclin E1 | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin, Protein Array | |
CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1 | |
Recombinant human Cyclin E1 protein fragment (around aa 10-176) (exact sequence is proprietary) | |
20 μg | |
Cancer, Cell Cycle and Replication, Cellular Markers, Ovarian Carcinoma Cell Markers, Phospho Specific | |
898 | |
Store at 4°C. | |
IgG2b κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array | |
CCNE1/2460 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title