Learn More
Cyclin E1 Mouse anti-Human, Mouse, Alexa Fluor 700, Clone: 5F8C5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237247AF700
Catalog No. NBP237247X
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Cyclin E1
5F8C5
Western Blot, Flow Cytometry, ELISA
Purified
CCNE1
Purified recombinant fragment of human CCNE1 (AA: 307-410) expressed in E. Coli.
Protein A or G purified
Cancer, Cell Cycle and Replication, Cellular Markers, Ovarian Carcinoma Cell Markers, Phospho Specific
Primary
898
ELISA, Flow Cytometry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse