Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclobenzaprine Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog Number 18601939
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
6202-23-9
C20H22ClN
311.85
MFCD00079039
VXEAYBOGHINOKW-UHFFFAOYSA-N
hydrogen dimethyl(3-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}propyl)amine chloride
[H+].[Cl-].CN(C)CCC=C1C2=CC=CC=C2C=CC2=CC=CC=C12
6202-23-9
C20H22ClN
311.85
MFCD00079039
VXEAYBOGHINOKW-UHFFFAOYSA-N
hydrogen dimethyl(3-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}propyl)amine chloride
[H+].[Cl-].CN(C)CCC=C1C2=CC=CC=C2C=CC2=CC=CC=C12

C1570, 6202-23-9, C20H21N•HCl

Spectrum™ Chemical Cyclobenzaprine Hydrochloride, USP is a muscle relaxant used to treat skeletal muscle conditions such as pain or injury. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
UN Number UN2811
Quantity 25 g
Percent Purity 98 to 102%
Grade USP
Ignition Residue 0.1%
Loss on Drying 1%
Packaging Amber Glass Bottle
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit