Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclobenzaprine Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
6202-23-9
C20H22ClN
311.85
VXEAYBOGHINOKW-UHFFFAOYSA-N
hydrogen dimethyl(3-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}propyl)amine chloride
[H+].[Cl-].CN(C)CCC=C1C2=CC=CC=C2C=CC2=CC=CC=C12
6202-23-9
C20H22ClN
311.85
VXEAYBOGHINOKW-UHFFFAOYSA-N
hydrogen dimethyl(3-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}propyl)amine chloride
[H+].[Cl-].CN(C)CCC=C1C2=CC=CC=C2C=CC2=CC=CC=C12

C1570, 6202-23-9, C20H21N•HCl

Spectrum™ Cyclobenzaprine Hydrochloride, USP is a muscle relaxant used to treat skeletal muscle conditions such as pain or injury. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
UN Number UN2811
Ignition Residue 0.001
Percent Purity 98 to 102%
Loss on Drying 0.01
Packaging Amber Glass Bottle
Quantity 100 g