Alfa Aesar™ Cyclobutanecarbonitrile, 97%

Manufacturer: Alfa Aesar™ A1030922

Quantity 100g

 View more versions of this product

Catalog No. AAA1030922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4426-11-3
    Molecular Formula C5H7N
    Formula Weight 81.12
    MDL Number MFCD00013735
    UN Number UN1992
    Beilstein 2037573
    Synonym Cyanocyclobutane; Cyclobutyl cyanide
    Chemical Name or Material Cyclobutanecarbonitrile

    Specifications

    Formula Weight 81.12
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym Cyanocyclobutane; Cyclobutyl cyanide
    Health Hazard 2 GHS H Statement
    H331-H226-H302-H312
    Toxic if inhaled.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Flash Point 42°C (107°F)
    Molecular Formula C5H7N
    MDL Number MFCD00013735
    UN Number UN1992
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
    Boiling Point 144° to 146°C
    Beilstein 2037573
    Health Hazard 1 Danger
    Odor Recognizable
    CAS 4426-11-3
    Quantity 100g
    Chemical Name or Material Cyclobutanecarbonitrile
    Assay 97%

    Description

    Applications
    It is used for research or analysis use only, as pharmaceutical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents, acids.

    GHS H Statement
    H331-H226-H302-H312
    Toxic if inhaled.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P210-P261-P280-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit