Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Cyclobutanemethanol, 99%
Click to view available options
Quantity:
25g
100g
Specifications
Refractive Index | 1.4465 |
Density | 0.913 |
Quantity | 100g |
Flash Point | 39°C (102°F) |
Boiling Point | 143°C to 144°C |
UN Number | UN1105 |
Beilstein | 2036027 |
Formula Weight | 86.13 |
Percent Purity | 99% |
Chemical Name or Material | Cyclobutanemethanol |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS
EINECSNumber : 224-575-4
TSCA : No
Recommended Storage : Ambient temperatures