Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclobutanemethanol, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
4415-82-1
C5H10O
86.13
MFCD00001330
WPOPOPFNZYPKAV-UHFFFAOYSA-N
cyclobutanemethanol,hydroxymethylcyclobutane,cyclobutylcarbinol,hydroxymethyl cyclobutane,cyclobutyl methanol,cyclobutyl-methanol,cyclobutanecarbinol,cyclobutane carbinol,pubchem23465
78130
cyclobutylmethanol
C1CC(C1)CO
4415-82-1
C5H10O
86.13
MFCD00001330
WPOPOPFNZYPKAV-UHFFFAOYSA-N
cyclobutanemethanol,hydroxymethylcyclobutane,cyclobutylcarbinol,hydroxymethyl cyclobutane,cyclobutyl methanol,cyclobutyl-methanol,cyclobutanecarbinol,cyclobutane carbinol,pubchem23465
78130
cyclobutylmethanol
C1CC(C1)CO
Specific Gravity 0.91
Density 0.9100g/mL
Flash Point 40°C
Boiling Point 143°C to 144°C
Refractive Index 1.4450 to 1.447
Packaging Glass bottle
Color Colorless
Infrared Spectrum Authentic
Quantity 5g
Assay Percent Range 99%
Linear Formula C4H7CH2OH
Solubility Information Solubility in water: immiscible
Formula Weight 86.13
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Cyclobutanemethanol, 99%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-575-4