Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclobutanol, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Has synthetic applications of gold-catalyzed ring expansions. Development of novel type of transition metal-catalyzed cascade reactions utilizing small ring systems and the applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Has synthetic applications of gold-catalyzed ring expansions. Development of novel type of transition metal-catalyzed cascade reactions utilizing small ring systems and the applications.
Solubility
Partly soluble.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid chlorides, acid anhydrides.
Has synthetic applications of gold-catalyzed ring expansions. Development of novel type of transition metal-catalyzed cascade reactions utilizing small ring systems and the applications.
Solubility
Partly soluble.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid chlorides, acid anhydrides.
Specifications
Density | 0.925 |
Boiling Point | 123°C to 124°C |
Flash Point | 21°C (69°F) |
Refractive Index | 1.436 |
Quantity | 1 g |
UN Number | UN1120 |
Beilstein | 2035937 |
Solubility Information | Partly soluble. |
Formula Weight | 72.11 |
Percent Purity | 96% |
Show More |
Safety and Handling
Hazard Category | H225 |
Hazard Statement | GHS H Statement H225 Highly flammable liquid and vapor. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTANOLS |
EINECSNumber | 220-858-1 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction