Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclobutanone, 98+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
1191-95-3
C4H6O
70.09
SHQSVMDWKBRBGB-UHFFFAOYSA-N
unii-6pf2sh405u,cyclobutyloxy,cyclobutanone, 98+%,cyclobutanon,cylcobutanone,cylobutanone,3-cyclobutanone,1-oxocyclobutane,cyclobutanone,pubchem9021
14496
cyclobutanone
C1CC(=O)C1
1191-95-3
C4H6O
70.09
SHQSVMDWKBRBGB-UHFFFAOYSA-N
unii-6pf2sh405u,cyclobutyloxy,cyclobutanone, 98+%,cyclobutanon,cylcobutanone,cylobutanone,3-cyclobutanone,1-oxocyclobutane,cyclobutanone,pubchem9021
14496
cyclobutanone
C1CC(=O)C1
Specific Gravity 0.93
Refractive Index 1.4205 to 1.4225
Melting Point -50.9°C
Flash Point 10°C
Boiling Point 99.0°C
Density 0.9300g/mL
Infrared Spectrum Authentic
Quantity 1g
Color Colorless to Yellow
Packaging Glass bottle
Assay Percent Range 98% min. (GC)
Linear Formula C4H6(=O)
Beilstein 07,5
Solubility Information Solubility in water: insoluble
Formula Weight 70.09
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material Cyclobutanone
Show More Show Less

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 214-745-6