Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclobutyl-4-fluorophenyl Ketone 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass Bottle
31431-13-7
C11H11FO
178.206
MFCD00019264
KMWTUCKAZFNWNG-UHFFFAOYSA-N
cyclobutyl 4-fluorophenyl ketone,cyclobutyl 4-fluorophenyl methanone,4-fluorobenzoylcyclobutane,cyclobutyl-4-fluorophenyl methanone,methanone,cyclobutyl 4-fluorophenyl
97471
cyclobutyl-(4-fluorophenyl)methanone
C1CC(C1)C(=O)C2=CC=C(C=C2)F
31431-13-7
C11H11FO
178.206
MFCD00019264
KMWTUCKAZFNWNG-UHFFFAOYSA-N
cyclobutyl 4-fluorophenyl ketone,cyclobutyl 4-fluorophenyl methanone,4-fluorobenzoylcyclobutane,cyclobutyl-4-fluorophenyl methanone,methanone,cyclobutyl 4-fluorophenyl
97471
cyclobutyl-(4-fluorophenyl)methanone
C1CC(C1)C(=O)C2=CC=C(C=C2)F
Refractive Index 1.5265 to 1.5285
Quantity 5g
Color Yellow
Flash Point 104°C
Boiling Point 125.0°C to 127.0°C (8.0 mmHg)
Packaging Glass Bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula FC6H4COC4H7
Formula Weight 178.21
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Cyclobutyl-4-fluorophenyl ketone, 98%
Show More Show Less