Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclobutyl methyl ketone, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
3019-25-8
C6H10O
98.145
MFCD00037148
JPJOOTWNILDNAW-UHFFFAOYSA-N
cyclobutyl methyl ketone,1-cyclobutylethan-1-one,acetylcyclobutane,ethanone, 1-cyclobutyl,ketone, cyclobutyl methyl,methyl cyclobutyl ketone,cyclobutylmethylketone,acetylcyclobutan,cyclobutylethanone,cyclobutane, acetyl
76398
1-cyclobutylethanone
CC(=O)C1CCC1
3019-25-8
C6H10O
98.145
MFCD00037148
JPJOOTWNILDNAW-UHFFFAOYSA-N
cyclobutyl methyl ketone,1-cyclobutylethan-1-one,acetylcyclobutane,ethanone, 1-cyclobutyl,ketone, cyclobutyl methyl,methyl cyclobutyl ketone,cyclobutylmethylketone,acetylcyclobutan,cyclobutylethanone,cyclobutane, acetyl
76398
1-cyclobutylethanone
CC(=O)C1CCC1
Packaging Glass bottle
Color Colorless to Green-Yellow
Density 0.9020g/mL
Quantity 5g
Infrared Spectrum Authentic
Refractive Index 1.4310 to 1.4330 (20°C, 589nm)
Assay Percent Range 98%
Linear Formula C4H7COCH3
Beilstein 07, IV, 37
Formula Weight 98.14
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Cyclobutyl methyl ketone
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning