Alfa Aesar™ Cyclobutyl phenyl ketone, 97%

Manufacturer: Alfa Aesar™ B2514603

Quantity 1g

 View more versions of this product

Catalog No. AAB2514603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5407-98-7
    Molecular Formula C11H12O
    Formula Weight 160.22
    MDL Number MFCD00001326
    Beilstein 2044307
    Synonym Benzoylcyclobutane
    Chemical Name or Material Cyclobutyl phenyl ketone

    Specifications

    Formula Weight 160.22
    Molecular Formula C11H12O
    MDL Number MFCD00001326
    CAS 5407-98-7
    Beilstein 2044307
    Synonym Benzoylcyclobutane
    Flash Point >110°C (230°F)
    Boiling Point 114° to 118°C (7mmHg)
    Quantity 1g
    Chemical Name or Material Cyclobutyl phenyl ketone
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit