Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cyclobutyl phenyl ketone, 97%

Manufacturer:  Alfa Aesar™ B2514614

Quantity 25g

 View more versions of this product

Catalog No. AAB2514614

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

5407-98-7
160.22
2044307
Cyclobutyl phenyl ketone
C11H12O
MFCD00001326
Benzoylcyclobutane

Specifications

>110°C (230°F)
25g
C11H12O
MFCD00001326
Benzoylcyclobutane
97%
114° to 118°C (7mmHg)
5407-98-7
160.22
2044307
Cyclobutyl phenyl ketone
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit