Alfa Aesar™ Cyclobutylamine hydrochloride, 97%

Manufacturer: Alfa Aesar™ B2540006

Quantity 5g

 View more versions of this product

Catalog No. AAB2540006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6291-01-6
    Molecular Formula C4H9N·HCl
    Formula Weight 107.58
    MDL Number MFCD00034953
    Synonym Aminocyclobutane hydrochloride
    Chemical Name or Material Cyclobutylamine hydrochloride

    Specifications

    Formula Weight 107.58
    Molecular Formula C4H9N·HCl
    MDL Number MFCD00034953
    CAS 6291-01-6
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 180° to 183°C
    Quantity 5g
    Synonym Aminocyclobutane hydrochloride
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Cyclobutylamine hydrochloride
    Assay 97%

    Description

    Applications
    Cyclobutylamine hydrochloride was used in the preparation of cyclobutylarachidonylamide.

    Solubility
    Soluble in water.

    Notes
    Hygroscopic. Store in cool, dry conditions, in well sealed container. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit