Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cyclobutylamine hydrochloride, 97%

Click to view available options
Quantity:
5g
25g
6291-01-6
C4H10ClN
107.58
MFCD00034953
NFAZOGXQOWEWBM-UHFFFAOYSA-N
cyclobutanamine hydrochloride,cyclobutylamine hydrochloride,aminocyclobutane hcl,cyclobutanamine, hydrochloride,cyclobutanaminium chloride,cyclobutylamine hcl,acmc-209wdt,aminocyclobutane hydrochloride,cyclobutylamine hydrochloric acid,cyclobutylamine hydrochloride salt
12618374
cyclobutanamine;hydrochloride
C1CC(C1)N.Cl
6291-01-6
C4H10ClN
107.58
MFCD00034953
NFAZOGXQOWEWBM-UHFFFAOYSA-N
cyclobutanamine hydrochloride,cyclobutylamine hydrochloride,aminocyclobutane hcl,cyclobutanamine, hydrochloride,cyclobutanaminium chloride,cyclobutylamine hcl,acmc-209wdt,aminocyclobutane hydrochloride,cyclobutylamine hydrochloric acid,cyclobutylamine hydrochloride salt
12618374
cyclobutanamine;hydrochloride
C1CC(C1)N.Cl
Applications
Cyclobutylamine hydrochloride was used in the preparation of cyclobutylarachidonylamide.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool, dry conditions, in well sealed container. Incompatible with oxidizing agents.
Quantity 25g
Melting Point 180°C to 183°C
Formula Weight 107.58
Percent Purity 97%
Chemical Name or Material Cyclobutylamine hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning