Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cyclododecanol, 99%

Click to view available options
Quantity:
25g
500g
1724-39-6
C12H24O
184.323
MFCD00003717
SFVWPXMPRCIVOK-UHFFFAOYSA-N
unii-0z645y4jts,dsstox_cid_7429,dsstox_rid_78451,dsstox_gsid_27429,acmc-1bprz
15595
cyclododecanol
C1CCCCCC(CCCCC1)O
1724-39-6
C12H24O
184.323
MFCD00003717
SFVWPXMPRCIVOK-UHFFFAOYSA-N
unii-0z645y4jts,dsstox_cid_7429,dsstox_rid_78451,dsstox_gsid_27429,acmc-1bprz
15595
cyclododecanol
C1CCCCCC(CCCCC1)O
Melting Point 77°C to 79°C
Quantity 500g
Flash Point 138°C (280°F)
UN Number UN3077
Formula Weight 184.32
Percent Purity 99%
Chemical Name or Material Cyclododecanol

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : TSCA