Alfa Aesar™ Cyclododecanol, 99%

Manufacturer: Alfa Aesar™ B2348622

Quantity 100g

 View more versions of this product

Catalog No. AAB2348622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1724-39-6
    Molecular Formula C12H24O
    Formula Weight 184.32
    MDL Number MFCD00003717
    UN Number UN3077
    Synonym Hydroxycyclododecane
    Chemical Name or Material Cyclododecanol

    Specifications

    Formula Weight 184.32
    TSCA TSCA
    Molecular Formula C12H24O
    MDL Number MFCD00003717
    CAS 1724-39-6
    UN Number UN3077
    Synonym Hydroxycyclododecane
    Melting Point 77° to 79°C
    Quantity 100g
    Flash Point 138°C (280°F)
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Chemical Name or Material Cyclododecanol
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit