Alfa Aesar™ Cyclododecene, 97%

Manufacturer: Alfa Aesar™ 4470930

Quantity 250g

 View more versions of this product

Catalog No. AA4470930

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1501-82-2
    Molecular Formula C12H22
    Formula Weight 160.31
    MDL Number MFCD00003721
    Chemical Name or Material Cyclododecene

    Specifications

    Formula Weight 160.31
    CAS 1501-82-2
    Physical Form Liquid
    Molecular Formula C12H22
    MDL Number MFCD00003721
    Refractive Index 1.4882
    Density 0.863g/mL
    Boiling Point 240°C
    Flash Point 93°C (199°F)
    Quantity 250g
    Chemical Name or Material Cyclododecene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit