missing translation for 'onlineSavingsMsg'
Learn More

Cyclofenil, Tocris Bioscience™

Catalog Number 399950
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
2624-43-3
C23H24O4
364.441
GVOUFPWUYJWQSK-UHFFFAOYSA-N
cyclofenil, ondogyne, cyclofenyl, cyclopenil, cyclophenyl, fertodur, cyclophenil, sanocrisin, ondonid, rehibin
2898
[4-[(4-acetyloxyphenyl)-cyclohexylidenemethyl]phenyl] acetate
CC(=O)OC1=CC=C(C=C1)C(=C2CCCCC2)C3=CC=C(C=C3)OC(=O)C
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
2624-43-3
C23H24O4
364.441
GVOUFPWUYJWQSK-UHFFFAOYSA-N
cyclofenil, ondogyne, cyclofenyl, cyclopenil, cyclophenyl, fertodur, cyclophenil, sanocrisin, ondonid, rehibin
2898
[4-[(4-acetyloxyphenyl)-cyclohexylidenemethyl]phenyl] acetate
CC(=O)OC1=CC=C(C=C1)C(=C2CCCCC2)C3=CC=C(C=C3)OC(=O)C

Selective estrogen receptor modulator (SERM)

Non-steroidal selective estrogen receptor modulator (SERM). Displays higher affinity for estrogen receptor β (ERβ) than for ERα.
Quantity 50 mg
Formula Weight 364.43
Percent Purity >99%
Chemical Name or Material Cyclofenil
Recommended Storage Store at +4°C
missing translation for 'provideContentCorrection'

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

missing translation for 'productTitle'