Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cycloheptane 98.0+%, TCI America™

Click to view available options
Quantity:
5mL
25mL
291-64-5
C7H14
98.19
MFCD00004147
DMEGYFMYUHOHGS-UHFFFAOYSA-N
heptamethylene, unii-vtz53p34ja, vtz53p34ja, cycloheptan, hsdb 59, cycloheptane, un2241 flammable liquid
9265
cycloheptane
C1CCCCCC1
291-64-5
C7H14
98.19
MFCD00004147
DMEGYFMYUHOHGS-UHFFFAOYSA-N
heptamethylene, unii-vtz53p34ja, vtz53p34ja, cycloheptan, hsdb 59, cycloheptane, un2241 flammable liquid
9265
cycloheptane
C1CCCCCC1
Color Colorless
Boiling Point 118°C
Quantity 25mL
UN Number 2241
Formula Weight 98.19
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material Cycloheptane