Alfa Aesar™ Cycloheptene, 96%, stab. with 0.1% BHT

Manufacturer: Alfa Aesar™ H2906214

Quantity 25g

 View more versions of this product

Catalog No. AAH2906214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 628-92-2
    Molecular Formula C7H12
    Formula Weight 96.17
    MDL Number MFCD00004156
    UN Number UN2242
    Chemical Name or Material Cycloheptene, stabilized with 0.1% BHT

    Specifications

    Formula Weight 96.17
    Molecular Formula C7H12
    Health Hazard 2 GHS H Statement
    H225
    Highly flammable liquid and vapor.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CYCLOHEPTENE
    Boiling Point 112° to 115°C
    Quantity 25g
    MDL Number MFCD00004156
    CAS 628-92-2
    UN Number UN2242
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point −7°C (19°F)
    Chemical Name or Material Cycloheptene, stabilized with 0.1% BHT
    Assay 96%

    Description

    Applications
    Cycloheptene was used in one-pot synthesis of 1,2/3-triols from the allylichydroperoxides catalyzed by zeolite-confined osmium(0) nanoclusters. It is a raw material in organic chemistry and a monomer in polymer synthesis. Copper (1) triflate catalyzes the photodimerization of the simple nonconjugated unstrained olefins, cyclopentene, cyclohexene, and cycloheptene, but not cyclooctene or acyclic olefins.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids. May react exothermically with reducing agents to release hydrogen gas.

    GHS H Statement
    H225
    Highly flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit