Alfa Aesar™ (Cyclohexadiene)iron tricarbonyl, 97+%

Manufacturer: Alfa Aesar™ 3940304

Quantity 2g

 View more versions of this product

Catalog No. AA3940304

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12152-72-6
    Formula Weight 220.01
    MDL Number MFCD00070435
    UN Number UN3281
    Synonym Tricarbonylcyclohexadiene iron
    Chemical Name or Material (Cyclohexadiene)iron tricarbonyl

    Specifications

    Formula Weight 220.01
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00070435
    UN Number UN3281
    Synonym Tricarbonylcyclohexadiene iron
    Health Hazard 1 Danger
    Physical Form Liquid
    CAS 12152-72-6
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, LIQUID, N.O.S.
    Sensitivity Air Sensitive
    Melting Point 8°C
    Health Hazard 2 GHS H Statement
    H300-H330
    Fatal if swallowed.
    Fatal if inhaled.
    Quantity 2g
    Chemical Name or Material (Cyclohexadiene)iron tricarbonyl
    Assay ≥97%

    GHS H Statement
    H300-H330
    Fatal if swallowed.
    Fatal if inhaled.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit