Learn More
Cyclohexane, ACS, 99+%, Thermo Scientific Chemicals
Description
Cyclohexane is used in environmental and GC analysis; for industrial production of adipic acid and caprolactam, which is a precursor of nylon and as a solvent, and to make nylon and paint remover.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 6.5°C |
Color | Colorless |
Appearance | Colorless liquid |
Density | 0.779 g/cm3 |
Boiling Point | 80°C to 81°C |
Quantity | 500 mL |
Flash Point | -18°C (-1°F) |
Ignition Point | 245°C (473°F) |
Odor | Pungent |
Physical Form | Liquid |
Show More |
Safety and Handling
Hazard Category | H225-H304-H315-H320-H336 |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Hazard Class: 3, Packing Group: II |
EINECSNumber | 203-806-2 |
RTECSNumber | GU6300000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.