Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
1L
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle

Chemical Identifiers

110-82-7
C6H12
84.162
XDTMQSROBMDMFD-UHFFFAOYSA-N
8078
CHEBI:29005
cyclohexane
C1CCCCC1

High quality chemicals for laboratory and specialized industrial use

Formula Weight 84.16
Packaging Narrow Mouth PVC Coated Amber Bottle
Specific Gravity 1L = 0.78kg
Flash Point −18.00°C
Quantity 4L
Chemical Name or Material Cyclohexane
Grade Baker Analyzed™ ACS Reagent