Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
500mL
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle

Chemical Identifiers

110-82-7
C6H12
84.162
XDTMQSROBMDMFD-UHFFFAOYSA-N
8078
CHEBI:29005

High quality chemicals for laboratory and specialized industrial use

CAS 110-82-7
Molecular Formula C6H12
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
SMILES C1CCCCC1
IUPAC Name cyclohexane
Molecular Weight (g/mol) 84.162
PubChem CID 8078
ChEBI CHEBI:29005
Formula Weight 84.16
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.78kg
Flash Point −18.00°C
Quantity 500mL
Chemical Name or Material Cyclohexane
Grade Baker Analyzed™ ACS Reagent
Show More Show Less